Sodium Bicarbonate, 1 M Buffer Solution, pH 8.0, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J62495-AP
AAJ62495-APEA
159.69
USD
AAJ62495-AP
Sodium Bicarbonate, 1 M Buffer Solution, pH 8.0, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 144-55-8
Physical Form: Liquid
MDL No.: MFCD00003528
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...