Polyvinylpyrrolidone 40,000 (K30), M.W. ∼ 40,000 (K30)

Supplier: Thermo Scientific Chemicals

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

J62417-36 J62417-A1 J62417-22
AAJ62417-36EA 223 USD
AAJ62417-36 AAJ62417-A1 AAJ62417-22
Polyvinylpyrrolidone 40,000 (K30), M.W. ∼ 40,000 (K30)
Polyvinylpyrrolidone

A cryoprotectant, also useful in plant cell culture research


Powder


A cryoprotectant, also useful in plant cell culture research

Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8
Merck Index: 14,7697

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR