Glycine ≥99.5% cell culture reagent
Supplier: Thermo Scientific Chemicals
Synonyms:
Aminoacetic acid, Glycocoll, H-Gly-OH
J62407-22
J62407-36
J62407-A1
AAJ62407-22EA
31.21
USD
AAJ62407-22
AAJ62407-36
AAJ62407-A1
Glycine ≥99.5% cell culture reagent
Glycine
Powder
Purity: 99.5+%
CAS No.: 56-40-6
Molecular Formula: C2H5NO2
Formula Weight: 75.07
Physical Form: Powder
Density: 1.595
Appearance: White to off-white
Odor: Odorless
pH: 5,97
Melting Point: ca 245° dec.
Beilstein Reference: 635782
Merck Reference: 14,4491
MDL No.: MFCD00008131
Formula:
NH₂CH₂COOH Boiling Pt: 181 °C (1013 hPa) Melting Pt: ∼ 245 °C (dec.) Density: 1.595 Storage Temperature: Ambient |
MDL Number:
MFCD00008131 CAS Number: 56-40-6 EINECS: 200-272-2 Merck Index: 14,4491 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...