Trichloroacetic acid (TCA) 10% (w/v) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: TCA, Trichloroethanoic acid, Aceto-Caustin, TCA

J62355-AP
AAJ62355-APEA 149.1 USD
AAJ62355-AP
Trichloroacetic acid (TCA) 10% (w/v) in aqueous solution
Trichloroacetic acid (TCA)

Protein precipitation reagent


CAS: 76-03-9 MDL: MFCD00004177

Formula: C₂HCl₃O₂
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: 54…56 °C
Density: Max. 1.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
UN: 2564
ADR: "8",III
Merck Index: 14,9627
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR