Rhodamine 6G

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 9-(2-(Ethoxycarbonyl)phenyl)-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride, 9-[2-(Ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride

J62315-14 J62315-22
AAJ62315-14EA 53.82 USD
AAJ62315-14 AAJ62315-22
Rhodamine 6G
Rhodamine 6G

Suitable for fluorescent labelling


Powder


Suitable for fluorescent labelling

Formula: C₂₈H₃₁ClN₂O₃
Boiling Pt: ? 200 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00012665
CAS Number: 989-38-8
EINECS: 213-584-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR