(R)-(-)-2-Oxothiazolidine-4-carboxylic acid
Supplier: Thermo Scientific Chemicals
|
Warning
|
J62254-03
AAJ62254-03EA
55.2
USD
AAJ62254-03
AAJ62254-06
(R)-(-)-2-Oxothiazolidine-4-carboxylic acid
(R)-(-)-2-Oxothiazolidine-4-carboxylic acid
Reagent for hepatic glutathione studies.
Powder
Reagent for hepatic glutathione studies.
Formula:
C₄H₅NO₃S Melting Pt: 174 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00066092 CAS Number: 19771-63-2 Merck Index: 14,6950 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...