Josamycin ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: (2S,3S,4R,6S)-6-(((2R,3S,4R,5R,6S)-6-(((4R,5S,6S,7R,9R,10R,11E,13E,16R)-4-Acetoxy-10-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)oxacyclohexadeca-11,13-dien-6-yl)oxy)-4-(dimethylamino)-5-hydroxy-2-methyltetrahydro-2H-pyran-3-yl)oxy)-4-hydroxy-2,4-

J62245-MF
AAJ62245-MFEA 112.22 USD
AAJ62245-MF
Josamycin ≥98%
Josamycin

Josamycin is used as a macrolide antibiotic effective against a variety of pathogens.


White to pale yellow powder


Applications: Macrolide antibiotic


Soluble in ethanol


Caution: Protect from light and moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Formula: C₄₂H₆₉NO₁₅
Storage Temperature: Refrigerator
MDL Number: MFCD00211043
CAS Number: 16846-24-5
EINECS: 240-871-6
Merck Index: 14,5267

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR