PIPES, 1.0 M Buffer Solutions, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60300-AK
J60300-AP
J60618-AK
J60618-AP
J60659-AK
J60659-AP
J62195-AK
J62195-AP
J62494-AK
J62494-AP
AAJ60300-AKEA
166.66
USD
AAJ60300-AK
AAJ60300-AP
AAJ60618-AK
AAJ60618-AP
AAJ60659-AK
AAJ60659-AP
AAJ62195-AK
AAJ62195-AP
AAJ62494-AK
AAJ62494-AP
PIPES, 1.0 M Buffer Solutions, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 5625-37-6
Appearance: Colorless
MDL No.: MFCD00006159
Caution: Storage Temperature: 2 to 8 °C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...