PIPES, 1.0 M Buffer Solutions, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J60300-AK J60300-AP J60618-AK J60618-AP J60659-AK J60659-AP J62195-AK J62195-AP J62494-AK J62494-AP
AAJ60300-AKEA 166.66 USD
AAJ60300-AK AAJ60300-AP AAJ60618-AK AAJ60618-AP AAJ60659-AK AAJ60659-AP AAJ62195-AK AAJ62195-AP AAJ62494-AK AAJ62494-AP
PIPES, 1.0 M Buffer Solutions, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 5625-37-6
Appearance: Colorless
MDL No.: MFCD00006159


Caution: Storage Temperature: 2 to 8 °C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR