Diosmin

Supplier: Thermo Scientific Chemicals

Synonyms: 5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one

J62073-06 J62073-14
AAJ62073-06EA 37.9 USD
AAJ62073-06 AAJ62073-14 AAJ62073-22
Diosmin
Diosmin

Natural flavanoid with anti-inflammatory effects


Powder


Natural flavanoid with anti-inflammatory effects

Formula: C₂₈H₃₂O₁₅
MW: 608.5 g/mol
Melting Pt: 274 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009772
CAS Number: 520-27-4
EINECS: 208-289-7
Merck Index: 14,3297

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR