Sodium cholate

Supplier: Thermo Scientific Chemicals

Synonyms: 3α,7α,12α-Trihydroxy-5β-cholanic acid sodium salt, 3α,7α,12α-Trihydroxycholan-24-oic acid sodium salt, 3α,7α,12α-Trihydroxycholanic acid sodium salt, Sodium 3α,7α,12α-trihydroxy-5β-cholanoate, (R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid sodium salt, 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid sodium salt

J62050-14 J62050-22 J62050-30
AAJ62050-14EA 58.2 USD
AAJ62050-14 AAJ62050-22 AAJ62050-30
Sodium cholate
Sodium cholate

Biochemical solubilizing agent


Powder


Biochemical solubilizing agent

Formula: C₂₄H₃₉NaO₅
MW: 430.56 g/mol
Melting Pt: 198
Storage Temperature: Ambient
MDL Number: MFCD00150749
CAS Number: 361-09-1
EINECS: 206-643-5
Merck Index: 14,2203

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR