Lansoprazole 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole, 2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulfinyl)-1H-benzo[d]imidazole

J62008-03 J62008-06 J62008-ME
AAJ62008-03EA 130 USD
AAJ62008-03 AAJ62008-06 AAJ62008-ME
Lansoprazole 98+%
Lansoprazole

A proton pump inhibitor


Crystalline powder


A proton pump inhibitor

Formula: C₁₆H₁₄F₃N₃O₂S
MW: 369.36 g/mol
Melting Pt: 173…175 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00866873
CAS Number: 103577-45-3
Merck Index: 14,5362

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR