Inosine 5'-monophosphate disodium salt hydrate

Supplier: Thermo Scientific Chemicals

Synonyms: 5'-IMP 2Na hydrate, 5'-Inosinic acid disodium salt hydrate, Inosinic acid (disodium)(hydrate)(1:2:X), Inosine 5'-monophosphate disodium salt xhydrate, Disodium inosinate hydrate, Inosic acid disodium salt hydrate

J61959-06 J61959-14 J61959-22
AAJ61959-06EA 37.99 USD
AAJ61959-06 AAJ61959-14 AAJ61959-22
Inosine 5'-monophosphate disodium salt hydrate
Inosine 5'-monophosphate disodium salt hydrate

Crystalline powder


CAS No.: 4691-65-0 Molecular Formula: C10H11N4Na2O8P.xH2O Formula Weight: 392.17(anhy) Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Appearance: White MDL No.: MFCD00036201

Formula: C₁₀H₁₁N₄Na₂O₈P·xH₂O
MW: 392.17 g/mol (anhy)
Storage Temperature: Refrigerator
MDL Number: MFCD00150372
CAS Number: 352195-40-5
EINECS: 225-146-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR