Terfenadine

Supplier: Thermo Scientific Chemicals

J61936-MC
AAJ61936-MCEA 288.5 USD
AAJ61936-MC
Terfenadine
Terfenadine

An antihistamine


Powder


Applications: An antihistamine

Formula: C₃₂H₄₁NO₂
Melting Pt: 145…152 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00079622
CAS Number: 50679-08-8
EINECS: 256-710-8
Merck Index: 14,9163

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR