Luzindole 97%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Acetyl-2-benzyl-tryptamine

J61915-LB0
AAJ61915-LB0EA 138 USD
AAJ61915-LB0 AAJ61915-MB
Luzindole 97%
Luzindole

A melatonin receptor antagonist


Powder


A melatonin receptor antagonist

Formula: C₁₉H₂₀N₂O
MW: 292.382 g/mol
Melting Pt: 44…46 °C
Storage Temperature: Freezer
MDL Number: MFCD00672498
CAS Number: 117946-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR