Blasticidin S hydrochloride 10 mg/ml in HEPES (1M)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Blasticidin S hydrochloride is used to inhibit protein synthesis in prokaryotic and eukaryotic organisms. It also induces apoptosis.
- Soluble in water
Purity: 98+%
CAS No.: 3513-03-9
Molecular Formula: C17H26N8O5.HCl
Formula Weight: 458.90
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
Merck Reference: 14,1316
UN No.: 2810
MDL No.: MFCD02091640
Inhibits protein synthesis and induces apoptosis
Caution: Store in cool place. Recommended to prepare and use solutions on the same day. Incompatible with strong oxidizing agents and strong bases.
Formula:
C₁₇H₂₇ClN₈O₅ Storage Temperature: Freezer |
MDL Number:
MFCD02091640 CAS Number: 3513-03-9 UN: 2810 ADR: 6.1,III Merck Index: 14,1316 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...