Sodium chloride 5 M in aqueous solution, autoclaved pH 8,0
Supplier: Thermo Scientific Chemicals
J61807-AK
AAJ61807-AKEA
98.18
USD
AAJ61807-AK
Sodium chloride 5 M in aqueous solution, autoclaved pH 8,0
Sodium chloride
Liquid
CAS No.: 7647-14-5
Molecular Formula: ClNa
Formula Weight: 58.44
Physical Form: Liquid
MDL No.: MFCD00003477
Formula:
NaCl Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...