Minoxidil

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,6-Diamino-4-(piperidin-1-yl)pyrimidine 1-oxide, Rogaine, Loniten

J61803-03 J61803-06 J61803-14
AAJ61803-03EA 31.96 USD
AAJ61803-03 AAJ61803-06 AAJ61803-14
Minoxidil
Minoxidil

Stimulates ATP-activated potassium channels


Crystalline powder


Applications: Stimulates ATP-activated potassium channels

Formula: C₉H₁₅N₅O
MW: 209.253 g/mol
Melting Pt: 248…265 °C
Storage Temperature: Ambient
MDL Number: MFCD00063409
CAS Number: 38304-91-5
EINECS: 253-874-2
Merck Index: 14,6203

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR