Acarbose 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Prandase, BAY G 5421, Acarbose Hydrate, 4",6"-Dideoxy-4"-([1S]-[1,4,6/5]-4,5,6-trihydroxy-3-hydroxymethyl-2-yclohexenylamino)-maltotriose, Precose, (2R,3R,4R,5R)-4-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4S,5S,6R)-3,4-Dihydroxy-6-methyl-5-(((1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-en-1-yl)amino)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal, Glucobay

J61737-03 J61737-04 J61737-06
AAJ61737-03EA 132 USD
AAJ61737-03 AAJ61737-04 AAJ61737-06
Acarbose 95%
Acarbose

A reversible alpha-glucosidase inhibitor


Powder


Applications: A reversible alpha-glucosidase inhibitor

Formula: C₂₅H₄₃NO₁₈
MW: 645.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00869592
CAS Number: 56180-94-0
EINECS: 260-030-7
Merck Index: 14,18

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR