Suramin hexasodium ≥98%

Supplier: Thermo Scientific Chemicals

J61707-MD
AAJ61707-03EA 1650.38 USD
AAJ61707-03 AAJ61707-ME AAJ61707-MD
Suramin hexasodium ≥98%
Suramin hexasodium

An antitumor and antiparasitic compound. Inhibits reverse transcriptase


Powder


Applications: An antitumor and antiparasitic compound. Inhibits reverse transcriptase

Formula: C₅₁H₃₄N₆Na₆O₂₃S₆
Storage Temperature: Refrigerator
MDL Number: MFCD00210217
CAS Number: 129-46-4
EINECS: 204-949-3
Merck Index: 14,9006

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR