Aniracetam

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(4-Methoxybenzoyl)pyrrolidin-2-one, 1-(4-Methoxybenzoyl)-2-pyrrolidone

J61661-03
AAJ61661-03EA 104 USD
AAJ61661-03
Aniracetam
Aniracetam

AMPA receptor potentiator. Slows the rate of ion-channel closing


Powder


Applications: AMPA receptor potentiator. Slows the rate of ion-channel closing

Formula: C₁₂H₁₃NO₃
MW: 219.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153767
CAS Number: 72432-10-1
Merck Index: 14,662

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR