Streptozocin 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Methyl-1-nitroso-3-((2S,3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)urea, Streptozotocin
Antibiotic and antitumor agent. Potent methylating agent for DNA
Purity: 97+%
CAS No.: 18883-66-4
Molecular Formula: C8H15N3O7
Formula Weight: 265.22
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White to Off-white
Melting Point: 121°
Solubility: Soluble in water or 100% ethanol
Beilstein Reference: 2060675
Merck Reference: 14,8832
MDL No.: MFCD00006607
Antibiotic and antitumor agent. Potent methylating agent for DNA
Formula:
C₈H₁₅N₃O₇ Melting Pt: 121 °C Storage Temperature: Freezer |
MDL Number:
MFCD00006607 CAS Number: 18883-66-4 EINECS: 242-646-8 Merck Index: 14,8832 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...