Emodin

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3,8-Trihydroxy-6-methylanthracene-9,10-dione, 1,3,8-Trihydroxy-6-methylanthraquinone

J61600-03 J61600-MC J61600-MD
AAJ61600-03EA 724 USD
AAJ61600-03 AAJ61600-MC AAJ61600-MD
Emodin
Emodin

A protein tyrosine kinase inhibitor


Powder


Applications: A protein tyrosine kinase inhibitor

Formula: C₁₅H₁₀O₅
Storage Temperature: Refrigerator
MDL Number: MFCD00001207
CAS Number: 518-82-1
EINECS: 208-258-8
Merck Index: 14,3561

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR