Sodium Phosphate, 0.5 M Buffer Solution, pH 7.5, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J61561-AK J61561-AP
AAJ61561-AKEA 119.41 USD
AAJ61561-AK AAJ61561-AP
Sodium Phosphate, 0.5 M Buffer Solution, pH 7.5, Thermo Scientific
Buffers Biological Buffers

Liquid


Physical Form: Liquid
MDL No.: MFCD00146206

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR