Gelatine Type A

Supplier: Thermo Scientific Chemicals

J61548-22 J61548-36 J61548-A1
AAJ61548-22EA 70.85 USD
AAJ61548-22 AAJ61548-36 AAJ61548-A1
Gelatine Type A
Proteins and Peptides

CAS No.: 9000-70-8
Physical Form: Powder
Appearance: Off-white to yellow
MDL No.: MFCD00081638

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR