4-Aminopyridine 99+%

Supplier: Thermo Scientific Chemicals
Danger

J61470-22 J61470-14
AAJ61470-22EA 177 USD
AAJ61470-22 AAJ61470-14
4-Aminopyridine 99+%
4-Aminopyridine

A potassium channel blocker


Crystalline powder


A potassium channel blocker

Formula: C₅H₆N₂
Boiling Pt: 273…274 °C
Melting Pt: 157…161 °C
Density: 1.26
Flash Pt: 164 °C(327 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006439
CAS Number: 504-24-5
EINECS: 207-987-9
UN: 2671
ADR: "6.1",II
Merck Index: 14,3933

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR