Polyvinylpyrrolidone 360,000 (K90), M.W. ∼ 360,000 (K90)
Supplier: Thermo Scientific Chemicals
Synonyms:
Povidone, Polyvinylpyrrolidon, Polyvidone, PVP
J61381-30
J61381-36
J61381-A1
AAJ61381-30EA
146.09
USD
AAJ61381-30
AAJ61381-36
AAJ61381-A1
Polyvinylpyrrolidone 360,000 (K90), M.W. ∼ 360,000 (K90)
Polyvinylpyrrolidone
A cryoprotectant, also useful in plant cell culture research
Powder
A cryoprotectant, also useful in plant cell culture research
Formula:
(C₆H₉NO)n Storage Temperature: Ambient |
MDL Number:
MFCD01076626 CAS Number: 9003-39-8 Merck Index: 14,7697 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...