Carboxy-PTIO potassium salt ≥98%
Supplier: Thermo Scientific Chemicals
J61316-MA J61316-MF
AAJ61316-MAEA 125 USD
AAJ61316-MA AAJ61316-MF
Carboxy-PTIO potassium salt ≥98%
Carboxy-PTIO potassium salt
Stable, water-soluble free radical that reacts with nitric oxide (NO) to inhibit NO synthase
Solid
Applications: Stable, water-soluble free radical that reacts with nitric oxide (NO) to inhibit NO synthase