4-Aminobutyric acid 99+%
Supplier: Thermo Scientific Chemicals
Powder
Purity: 99+%
CAS No.: 56-12-2
Molecular Formula: C4H9NO2
Formula Weight: 103.12
Physical Form: Powder
Appearance: White
Odor: Odorless
Melting Point: ca 195° dec.
Beilstein Reference: 906818
Merck Reference: 14,430
MDL No.: MFCD00008226
Formula:
C₄H₉NO₂ Boiling Pt: 248 °C (1013 hPa) Melting Pt: ∼ 195 °C (dec.) Density: 1.21 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008226 CAS Number: 56-12-2 EINECS: 200-258-6 Merck Index: 14,430 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...