Casein Blocking Buffer, 3% in TBS, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J61298-AP
AAJ61298-APEA
118
USD
AAJ61298-AP
Casein Blocking Buffer, 3% in TBS, Thermo Scientific
Buffers
Electrophoresis Buffers
Blocking Buffers
Liquid
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
MDL No.: MFCD01634587
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...