Kanamycin sulfate

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Kanamycin Monosulfate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulfate, Kanamycin A, Kanamycin Monosulphate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphate

J61272-09 J61272-14 J61272-22
AAJ61272-09EA 209.75 USD
AAJ61272-09 AAJ61272-14 AAJ61272-22
Kanamycin sulfate
Kanamycin sulphate

Antibiotic. Inhibits translocation during protein biosynthesis


Powder


Antibiotic. Inhibits translocation during protein biosynthesis

Formula: C₁₈H₃₈N₄O₁₅S
MW: 582.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070253
CAS Number: 25389-94-0
EINECS: 246-933-9
Merck Index: 14,5281

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR