Citrate, 0.5 M Buffer Solutions, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J60024-AK J60024-AP J60125-AK J60125-AP J60754-AK J60754-AP J61249-AK J61249-AP J61391-AK J61391-AP J61690-AK J61690-AP J63008-AK J63008-AP
AAJ60024-AKEA 171.65 USD
AAJ60024-AK AAJ60024-AP AAJ60125-AK AAJ60125-AP AAJ60754-AK AAJ60754-AP AAJ61249-AK AAJ61249-AP AAJ61391-AK AAJ61391-AP AAJ61690-AK AAJ61690-AP AAJ63008-AK AAJ63008-AP
Citrate, 0.5 M Buffer Solutions, Thermo Scientific
Buffers Biological Buffers

Colourless odourless buffer solutions.


  • CAS No.: 6132-04-3
  • MDL No.: MFCD00150031


Citrate, is used as an anticoagulant and also used as a biological buffer. It is often used to prepare sodium citrate buffer for antigen retrieval of tissue samples. The citrate solution is designed to break protein cross-links, thus, unmasking antigens and epitopes in formalin-fixed and paraffin embedded tissue sections, resulting in enhancing staining intensity of antibodies. Citrate has anticoagulant activity and as a calcium chelator, it forms complexes that disrupt the tendency of blood to clot.


Caution: Store in refrigerator. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR