TAPS sodium salt (3-(Tris(hydroxymethyl)methylamino)propane-1-sulfonic acid sodium salt) 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: TAPS sodium salt, Sodium 3-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)propane-1-sulfonate, Sodium 3-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)propane-1-sulphonate

J61189-14 J61189-22
AAJ61189-14EA 35.1 USD
AAJ61189-14 AAJ61189-22
TAPS sodium salt (3-(Tris(hydroxymethyl)methylamino)propane-1-sulfonic acid sodium salt) 98%
TAPS sodium salt (3-(Tris(hydroxymethyl)methylamino)propane-1-sulphonic acid sodium salt)

Zwitterionic buffer


Powder


Zwitterionic buffer

Formula: C₇H₁₆NNaO₆S
MW: 265.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00069903
CAS Number: 91000-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR