Zinc sulfate 100 mM in aqueous solution

Supplier: Thermo Scientific Chemicals
Warning

J60963-AE
AAJ60963-AEEA 77.61 USD
AAJ60963-AE
Zinc sulfate 100 mM in aqueous solution
Zinc sulphate

Liquid


CAS No.: 7733-02-0 Molecular Formula: O4SZn Formula Weight: 161.43 Physical Form: Liquid Appearance: Colorless UN No.: 3082 MDL No.: MFCD00011302

Formula: O₄SZn
MW: 161.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011302
CAS Number: 7733-02-0
EINECS: 231-793-3
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR