Potassium acetate, 1M aqua solution, pH 7.5, RNAse free, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60832-AK
AAJ60832-AEEA
65.7
USD
AAJ60832-AE
AAJ60832-AK
Potassium acetate, 1M aqua solution, pH 7.5, RNAse free, Thermo Scientific
Buffers
Cloning Buffers
CAS No.: 127-08-2
Molecular Formula: C₂H₃KO₂
Formula Weight: 98.15 g/mol
Physical Form: Liquid
Appearance: Colorless
Caution: RUO – Research Use Only
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...