1,3-Dihydroxyacetone dimer
Supplier: Thermo Scientific Chemicals
Powder
CAS No.: 62147-49-3
Molecular Formula: C3H6O3
Formula Weight: 180.16
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White to off-white
Melting Point: 75-80°
Merck Reference: 14,3182
MDL No.: MFCD00004670
Formula:
C₆H₁₂O₆ Melting Pt: 75…80 °C Density: 1.52 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00051019 CAS Number: 62147-49-3 EINECS: 202-494-5 Merck Index: 14,3182 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...