CHAPS Lysis Buffer, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60580-AD
J60580-AE
AAJ60580-ADEA
116.54
USD
AAJ60580-AD
AAJ60580-AE
CHAPS Lysis Buffer, Thermo Scientific
Buffers
Extraction Buffers
Liquid
Physical Form: Liquid
Appearance: Clear liquid
MDL No.: MFCD00282866
Applications: For cell lysis and protein extraction
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...