Polyvinylpyrrolidone 10,000 (K15), M.W. ∼ 10,000 (K15)
Supplier: Thermo Scientific Chemicals
Synonyms:
Povidone, Polyvinylpyrrolidon, Polyvidone, PVP
J60382-14
J60382-36
J60382-22
J60382-A1
AAJ60382-14EA
37.6
USD
AAJ60382-14
AAJ60382-36
AAJ60382-22
AAJ60382-A1
Polyvinylpyrrolidone 10,000 (K15), M.W. ∼ 10,000 (K15)
Polyvinylpyrrolidone
A cryoprotectant, also useful in plant cell culture research
Powder
A cryoprotectant, also useful in plant cell culture research
Formula:
(C₆H₉NO)n Storage Temperature: Ambient |
MDL Number:
MFCD01076626 CAS Number: 9003-39-8 Merck Index: 14,7697 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...