Polyvinylpyrrolidone 10,000 (K15), M.W. ∼ 10,000 (K15)

Supplier: Thermo Scientific Chemicals

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

J60382-14 J60382-36 J60382-22 J60382-A1
AAJ60382-14EA 37.6 USD
AAJ60382-14 AAJ60382-36 AAJ60382-22 AAJ60382-A1
Polyvinylpyrrolidone 10,000 (K15), M.W. ∼ 10,000 (K15)
Polyvinylpyrrolidone

A cryoprotectant, also useful in plant cell culture research


Powder


A cryoprotectant, also useful in plant cell culture research

Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8
Merck Index: 14,7697

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR