EDTA (ethylenediamine tetraacetic acid) 0.5 M in aqueous solution, autoclaved pH 8.0
Supplier: Thermo Scientific Chemicals
|
Warning
|
J60292-AK
J60292-AP
AAJ60292-AKEA
118.76
USD
AAJ60292-AK
AAJ60292-AP
EDTA (ethylenediamine tetraacetic acid) 0.5 M in aqueous solution, autoclaved pH 8.0
EDTA (Ethylenediamine tetraacetic acid)
Liquid
CAS No.: 60-00-4
Molecular Formula: C10H16N2O8
Formula Weight: 292.24
Physical Form: Liquid
MDL No.: MFCD00003541
Formula:
C₁₀H₁₆N₂O₈ Storage Temperature: Ambient |
MDL Number:
MFCD00003541 CAS Number: 60-00-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...