Nitro blue tetrazolium chloride (NBT) 99%

Supplier: Thermo Scientific Chemicals
Warning

J60230-03 J60230-06
AAJ60230-03EA 141.64 USD
AAJ60230-03 AAJ60230-06
Nitro blue tetrazolium chloride (NBT) 99%
Nitro blue tetrazolium chloride (NBT)

Intense blue dye used in histochemistry determinations


Crystalline powder


Intense blue dye used in histochemistry determinations

Formula: C₄₀H₃₀Cl₂N₁₀O₆
Melting Pt: ∼ 190 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012159
CAS Number: 298-83-9
EINECS: 206-067-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR