Theophyllin
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,3-Dimethyl-1H-purine-2,6(3H,9H)-dione, Theophylline
J60203-5H
AAJ60203-5HEA
118.26
USD
AAJ60203-5H
Theophyllin
Theophyllin
Phosphodiesterase inhibitor; diuretic; cardiac stimulant; muscle relaxant; asthma medication. A methylxanthine drug used in therapy for respiratory diseases such as COPD and asthma
Powder
Formula:
C₇H₈N₄O₂ MW: 180.16 g/mol Melting Pt: 270…274 °C Storage Temperature: Ambient |
MDL Number:
MFCD00079619 CAS Number: 58-55-9 EINECS: 200-385-7 UN: 2811 ADR: "6.1",III Merck Index: 14,9285 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...