Actinomycin D, powder

Supplier: Thermo Scientific Chemicals
Danger

J60148-LB0
AAJ60148-LB0EA 151.75 USD
AAJ60148-LB0
Actinomycin D, powder
Actinomycin D

Inhibits nulceic acid synthesis and potently induces apoptosis


  • Orange to red powder


Used in banding techniques to differentiate between different regions of chromosomes. Also used as a selection agent

Formula: C₆₂H₈₆N₁₂O₁₆
Melting Pt: 251…253 °C
Storage Temperature: Freezer
MDL Number: MFCD00005033
CAS Number: 50-76-0
EINECS: 200-063-6
UN: 3462
ADR: 6.1,II
Merck Index: 14,2800

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR