Acriflavine hydrochloride

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,6-Diamino-10-methylacridinium chloride hydrochloride, Euflavine

J60048-14
AAJ60048-14EA 52.4 USD
AAJ60048-14
Acriflavine hydrochloride
Acriflavine hydrochloride

An intercalating dye.


Powder

Formula: C₂₇H₂₈Cl₄N₆
MW: 578.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00069039
CAS Number: 8063-24-9
ADR: 9,III
Merck Index: 14,123

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR