Transformation Reagent, TB Solution-II, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J60028-AK
AAJ60028-AKEA 142 USD
AAJ60028-AK
Transformation Reagent, TB Solution-II, Thermo Scientific
Nucleic Acid Reagents Transformation Reagents
Used for the transoformation of competent cells.
Contains 250 mM potassium chloride, 15 mM calcium chloride, 10mM HEPES and 55 mM manganese chloride, pH 6.5.