Ethyl-7-nitroindole-2-carboxylate 95%

Supplier: Thermo Scientific Chemicals
Warning

H66657-03 H66657-06
AAH66657-03EA 90.1 USD
AAH66657-03 AAH66657-06
Ethyl-7-nitroindole-2-carboxylate 95%
Ethyl-7-nitroindole-2-carboxylate

Purity: 95% CAS No.: 6960-46-9 Molecular Formula: C11H10N2O4 Formula Weight: 234.21 Physical Form: Solid Beilstein Reference: 224930 MDL No.: MFCD00216475

Formula: C₁₁H₁₀N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00216475
CAS Number: 6960-46-9

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR