Fmoc-D-Bpa-OH 98%
Supplier: Thermo Scientific Chemicals
N-Fmoc-4-benzoyl-D-phenylalanine, 98%
Purity: 98%
CAS No.: 117666-97-4
Molecular Formula: C31H25NO5
Formula Weight: 491.54
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Pale brown
MDL No.: MFCD00237666
Formula:
C₃₁H₂₅NO₅ Storage Temperature: Ambient |
MDL Number:
MFCD00237666 CAS Number: 117666-97-4 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...