2-Amino-9,9-dimethylfluorene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64771-06
AAH64771-03EA
74.57
USD
AAH64771-03
AAH64771-06
AAH64771-MD
2-Amino-9,9-dimethylfluorene 98%
2-Amino-9,9-dimethylfluorene
2-Amino-9,9-dimethylfluorene, 98%
Purity: 98%
CAS No.: 108714-73-4
Molecular Formula: C15H15N
Formula Weight: 209.29
Physical Form: Powder
Appearance: Off-white
Beilstein Reference: 15506865
MDL No.: MFCD09953790
Formula:
C₁₅H₁₅N Storage Temperature: Ambient |
MDL Number:
MFCD09953790 CAS Number: 108714-73-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...