2-[2-(Trifluoromethyl)phenyl]ethylamine 95%

Supplier: Thermo Scientific Chemicals
Danger

H64492-03 H64492-06 H64492-14
AAH64492-03EA 53.2 USD
AAH64492-03 AAH64492-06 AAH64492-14
2-[2-(Trifluoromethyl)phenyl]ethylamine 95%
2-[2-(Trifluoromethyl)phenyl]ethylamine

2-[2-(Trifluoromethyl)phenyl]ethylamine, 95%


Purity: 95% CAS No.: 774-18-5 Molecular Formula: C9H10F3N Formula Weight: 189.18 Physical Form: Liquid Appearance: Pale yellow Beilstein Reference: 7202211 UN No.: 2735 MDL No.: MFCD00128232

Formula: C₉H₁₀F₃N
MW: 189.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00128232
CAS Number: 774-18-5
EINECS: 212-262-5
UN: 2735
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR