2-Chloro-5-nitropyrimidin-4-amine 97%
Supplier: Thermo Scientific Chemicals
H64478-03
H64478-06
AAH64478-03EA
80
USD
AAH64478-03
AAH64478-06
AAH64478-MD
2-Chloro-5-nitropyrimidin-4-amine 97%
2-Chloro-5-nitropyrimidin-4-amine
Purity: 97%
CAS No.: 1920-66-7
Molecular Formula: C4H3ClN4O2
Formula Weight: 174.54
Physical Form: Solid
Melting Point: 221-226°
Beilstein Reference: 156953
MDL No.: MFCD00127771
Formula:
C₄H₃ClN₄O₂ Melting Pt: 221…226 °C Storage Temperature: Ambient |
MDL Number:
MFCD00127771 CAS Number: 1920-66-7 EINECS: 217-648-7 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...